Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm13363 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002688.1 | 1 | 2414 |
Gene Symbol: Ptp4a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC094447.1 | - | 6 | 2328 | 3990529 |