Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: D730001G18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027836.1 | 3 | 1169 | |||
NR_027837.1 | 2 | 1070 | ||||
GenBank mRNA | AK021322.1 | - | 2 | 1071 | ||
AK037167.1 | - | 2 | 1070 | |||
BC055822.1 | - | 3 | 1134 | |||
BC147771.1 | - | 1 | 950 | |||
BC147774.1 | - | 1 | 950 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AA498853.1 | - | 1 | 394 | Chr. 15 - 74771262 - 74771727 [-] on Build GRCm38 |