Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: 2900052N01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015605.1 | 4 | 2051 | |||
GenBank mRNA | AK038437.1 | - | 5 | 1766 | ||
AK049847.1 | - | 4 | 2051 |