Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Neat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AK018202.1 | - | 1 | 63 | ||
AK028745.1 | - | LIT3353 VINC |
1 | 1699 | ||
AK033579.1 | - | 1 | 1697 | |||
AK145725.1 | - | 1 | 1697 | |||
AK148054.1 | - | 2 | 1531 | |||
AK159400.1 | - | 1 | 1696 | |||
AK159773.1 | - | 1 | 1699 | |||
AK167154.1 | - | 1 | 1698 | |||
AK167255.1 | - | 1 | 1697 | |||
BC025215.1 | - | 2 | 1052 | |||
BC064019.1 | - | 1 | 920 | |||
BC095948.1 | - | 1 | 877 | |||
EF177378.1 | - | 1 | 1696 | |||
GQ859163.1 | - | 1696 |