Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Neat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AK018202.1 | - | 1 | 910 | ||
AK028745.1 | - | LIT3353 VINC |
1 | 2546 | ||
AK033579.1 | - | 1 | 2544 | |||
AK145725.1 | - | 1 | 2544 | |||
AK159400.1 | - | 1 | 2543 | |||
AK159773.1 | - | 1 | 2546 | |||
AK167154.1 | - | 1 | 2545 | |||
AK167255.1 | - | 1 | 2544 | |||
BC025215.1 | - | 2 | 1899 | |||
BC064019.1 | - | 1 | 1767 | |||
BC095948.1 | - | 1 | 1724 | |||
EF177378.1 | - | 1 | 2543 | |||
GQ859163.1 | - | 2543 |