Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpina4-ps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002861.2 | 4 | 1158 | |||
GenBank mRNA | BC018155.1 | - | 4 | 1282 | ||
BC031891.1 | - | 4 | 3696 | |||
BC039883.1 | - | 1 | 300 |