Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700012D14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015573.2 | 1 | 53 | |||
GenBank mRNA | AK005898.1 | - | 1 | 55 | ||
AK031468.1 | - | 1 | 70 | |||
AK047116.1 | - | 1 | 70 | |||
AK087602.1 | - | 1 | 60 |