Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: F630028O10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_030718.1 | 3 | 650 | |||
GenBank mRNA | AK036748.1 | - | 2 | 1135 | ||
AK170206.1 | - | 3 | 635 |