Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1200015M12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK004796.1 | - | 1 | 1636 | ||
BC040222.1 | - | 1 | 2198 |
Gene Symbol: A130040M12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK037717.1 | - | 1066 | |||
AK149382.1 | - | 1 | 2671 | |||
AK153243.1 | - | 1 | 1288 |