Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtag2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015480.1 | 2 | 151 | |||
NR_027802.1 | 1 | 180 | ||||
GenBank mRNA | AK042966.1 | - | 2 | 151 | ||
BC107317.1 | - | 1 | 180 | |||
BC107318.1 | - | 1 | 180 | |||
BC119507.1 | - | 1 | 180 | |||
BC119508.1 | - | 1 | 180 | |||
BC127168.1 | - | 1 | 180 | |||
BC127169.1 | - | 1 | 180 | |||
U88401.1 | - | 1 | 179 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB640217.1 | - | 2 | 151 | Chr. 7 - 45366161 - 45367875 [+] on Build GRCm38 |