Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pea15b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY487415.1 | - | 169 | |||
BC132661.1 | - | 266 | 40130938 | |||
BC132663.1 | - | 266 | 40130940 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | NM_001010832.1 | - | 169 | Chr. - - [-] on Build |