Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C230004F18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_030706.1 | 6 | 542 | |||
GenBank mRNA | AK082088.1 | - | 523 | |||
AK163488.1 | - | 6 | 535 | |||
BB653348.1 | - | 395 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | NM_177916.1 | - | 523 | Chr. - - [-] on Build |