Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C4bp-ps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028304.1 | 1 | 55 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG296545.1 | - | 55 | Chr. - - [-] on Build |