Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLKP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002948.1 | 5 | 639 | |||
GenBank mRNA | AF355199.1 | - | 2 | 201 | ||
AF464754.1 | - | 2 | 377 | |||
AY533562.1 | - | 5 | 639 | |||
DQ086829.1 | - | 2 | 1046 | |||
DQ211698.1 | - | 5 | 2362 |