Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANP32AP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003144.1 | 1 | 285 | |||
GenBank mRNA | AK311417.1 | - | 2 | 487 | ||
DQ083510.1 | - | 1 | 285 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA693550.1 | - | 2 | 487 | Chr. 15 - 35181799 - 35237703 [+] on Build GRCh38 |