Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIPARP-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027954.1 | 1 | 2285 | |||
GenBank mRNA | BC033324.1 | - | 1 | 2251 | ||
BC038770.1 | - | 1 | 919 |