Gene Symbol: HECTD2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024467.1 | 4 | 469 | |||
GenBank mRNA | DA898704.1 | - | 4 | 469 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AA211636.1 | - | 2 | 141 | Chr. 10 - 91307263 - 91525125 [-] on Build GRCh38 |