Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100101148 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104007.1 | 4 | 419 | |||
GenBank mRNA | BC070189.1 | - | 4 | 419 |
Gene Symbol: LOC541473 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003602.3 | 5 | 557 | |||
GenBank mRNA | BC022013.2 | - | 5 | 511 | ||
BC067783.1 | - | 5 | 530 |