Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHKB-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_021492.2 | 2 | 403 | |||
GenBank mRNA | BE393070.1 | - | 2 | 304 | ||
BJ989458.1 | - | 2 | 284 | |||
CR456603.1 | - | 2 | 403 | |||
CR980518.1 | - | 2 | 307 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | HY124358.1 | - | 2 | 306 | Chr. 22 - 50583124 - 50583885 [+] on Build GRCh38 |