Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIR497 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_030178.1 | 1 | 75 |
Gene Symbol: LOC284112 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK098506.1 | - | 1 | 1712 |