Search Thermo Fisher Scientific
Gene Symbol: LINC00474 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024032.1 | 3 | 408 | |||
GenBank mRNA | AB021923.1 | - | 3 | 518 | ||
BC104240.1 | - | 3 | 408 | |||
BC104241.1 | - | 3 | 407 | |||
BC112996.1 | - | 3 | 408 | |||
BC112997.1 | - | 3 | 408 | |||
BC128039.1 | - | 3 | 408 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX116645.1 | - | 3 | 410 | Chr. 9 - 115888162 - 115925090 [-] on Build GRCh38 |