Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL32P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003111.2 | 4 | 1612 | |||
GenBank mRNA | BC015389.1 | - | 3 | 1176 | ||
BC053996.1 | - | 5 | 1783 |