Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_015342.2 | 3 | 1151 | |||
NR_132312.1 | 4 | 1316 | ||||
NR_132313.1 | 3 | 1016 | ||||
GenBank mRNA | AF103907.1 | - | LIT1552 DD3 |
4 | 1316 | |
AY894120.1 | - | 3 | 1149 |