Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM167A-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026814.1 | 6, 7 | 1009 | |||
GenBank mRNA | AJ301563.1 | - | 6, 7 | 1009 | ||
BC080558.1 | - | 6, 7 | 1143 |