Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XGY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003254.1 | 3 | 151 | |||
GenBank mRNA | Z48518.1 | - | 3 | 151 | ||
Z48519.1 | - | 3 | 151 |