Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00923 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024172.1 | 3 | 729 | |||
NR_024173.1 | 2 | 557 | ||||
GenBank mRNA | AK025311.1 | - | 3 | 850 | ||
BC105714.1 | - | 2 | 557 | |||
BC105715.1 | - | 3 | 729 | |||
BC105716.1 | - | 2 | 557 | |||
BC105717.1 | - | 2 | 557 |