Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMED5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_030761.1 | 2, 3 | 739 | |||
GenBank mRNA | AK125882.1 | - | 2, 3 | 439 | ||
BX642274.1 | - | 2, 3 | 452 | |||
CR936808.1 | - | 2, 3 | 467 |