Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMGXB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027780.1 | 4, 5 | 431 | |||
XM_006724102.1 | XP_006724165.1 | 3, 4 | 299 | |||
GenBank mRNA | AJ010070.1 | - | 4, 5 | 380 | ||
AK299833.1 | - | 4, 5 | 396 | |||
AL079310.1 | - | 4, 5 | 380 | |||
BC004194.2 | - | 1, 2 | 31 | |||
BC017326.2 | - | 3, 4 | 268 | |||
BC080611.1 | - | 4, 5 | 386 |