Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNORA11D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003711.1 | 1 | 105 |
Gene Symbol: MAGED4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011530823.2 | XP_011529125.1 | 11 | 2071 | ||
GenBank mRNA | AK094541.1 | - | 5 | 2092 |
Gene Symbol: SNORA11E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_102368.1 | 1 | 105 |