Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: LINC-PINT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015431.2 | 5 | 2135 | |||
NR_109850.1 | 6 | 2259 | ||||
NR_109851.1 | 6 | 2264 | ||||
NR_109852.1 | 6 | 2237 | ||||
GenBank mRNA | AK125651.1 | - | 6 | 2237 | ||
AL713664.1 | - | 4 | 952 | |||
BC047367.1 | - | 4 | 1281 | |||
BX451434.2 | - | 1 | 446 |