Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: LINC-PINT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015431.2 | 5 | 2665 | |||
NR_024153.2 | 5 | 801 | ||||
NR_109850.1 | 6 | 2789 | ||||
NR_109851.1 | 6 | 2794 | ||||
NR_109852.1 | 6 | 2767 | ||||
NR_109853.1 | 6 | 1835 | ||||
NR_109854.1 | 6 | 1791 | ||||
GenBank mRNA | AK125651.1 | - | 6 | 2767 | ||
AL713664.1 | - | 4 | 1482 | |||
BC039420.1 | - | 5 | 801 | |||
BC047367.1 | - | 4 | 1811 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI755625.1 | - | 5 | 678 | Chr. 7 - 130944567 - 131107827 [-] on Build GRCh38 |