Gene Symbol: ADAM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_001448.1 | 11, 12 | 1292 | |||
GenBank mRNA | AJ132821.1 | - | 10, 11 | 1226 | ||
AJ132822.1 | - | 10, 11 | 1108 | |||
AJ132823.1 | - | 9, 10 | 1042 | |||
AJ132827.1 | - | 1, 2 | 222 | |||
BC047448.1 | - | 11, 12 | 1426 | |||
BC067864.1 | - | 9, 10 | 1085 |