Gene Symbol: ADAM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_001448.1 | 13, 14 | 1491 | |||
GenBank mRNA | AJ132821.1 | - | 12, 13 | 1425 | ||
AJ132822.1 | - | 12, 13 | 1307 | |||
AJ132823.1 | - | 11, 12 | 1241 | |||
AJ132826.1 | - | 2, 3 | 334 | |||
BC047448.1 | - | 13, 14 | 1625 | |||
BC067864.1 | - | 11, 12 | 1284 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DB525201.1 | - | 1, 2 | 57 | Chr. 8 - 39393330 - 39417376 [+] on Build GRCh38 |