Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_001448.1 | 9 | 858 | |||
GenBank mRNA | AJ132821.1 | - | 8 | 792 | ||
AJ132822.1 | - | 8 | 674 | |||
AJ132823.1 | - | 7 | 608 | |||
BC047448.1 | - | 9 | 992 | |||
BC067864.1 | - | 7 | 651 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI560028.1 | - | 7 | 655 | Chr. 8 - 39314644 - 39375865 [+] on Build GRCh38 |