Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ESRRG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243515.1 | NP_001230444.1 | 3 | 500 | ||
NM_001243519.1 | NP_001230448.1 | 3 | 504 | |||
XM_011509267.1 | XP_011507569.1 | 3 | 457 | |||
XM_011509268.2 | XP_011507570.1 | 2 | 342 | |||
XM_011509276.1 | XP_011507578.1 | 3 | 493 | |||
XM_011509280.2 | XP_011507582.1 | 2 | 338 | |||
XM_017000626.1 | XP_016856115.1 | 2 | 271 | |||
XM_017000627.1 | XP_016856116.1 | 2 | 334 | |||
XM_017000628.1 | XP_016856117.1 | 3 | 457 | |||
XM_017000630.1 | XP_016856119.1 | 3 | 457 | |||
XM_017000635.1 | XP_016856124.1 | 3 | 457 | |||
GenBank mRNA | AK290945.1 | - | 3 | 481 | ||
BC008218.2 | - | 3 | 364 | |||
DA722064.1 | - | 3 | 456 |