Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD2BP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243646.1 | NP_001230575.1 | 1 | 151 | ||
GenBank mRNA | BC013854.1 | - | 1 | 151 |