Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BBIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195304.1 | NP_001182233.1 | 5 | 1072 | ||
NM_001195305.1 | NP_001182234.1 | 4 | 915 | |||
NM_001195306.1 | NP_001182235.1 | 4 | 761 | |||
NM_001195307.1 | NP_001182236.1 | 3 | 840 | |||
NM_001243783.1 | NP_001230712.1 | 3 | 822 | |||
GenBank mRNA | AK025724.1 | - | 4 | 735 | ||
AK307126.1 | - | 4 | 761 | |||
AK307314.1 | - | 5 | 948 | |||
BC105804.1 | - | 5 | 911 |