Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BBIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195304.1 | NP_001182233.1 | 5 | 1831 | ||
NM_001195305.1 | NP_001182234.1 | 4 | 1674 | |||
NM_001195306.1 | NP_001182235.1 | 4 | 1520 | |||
NM_001195307.1 | NP_001182236.1 | 3 | 1599 | |||
NM_001243783.1 | NP_001230712.1 | 3 | 1581 | |||
GenBank mRNA | AK025724.1 | - | 4 | 1494 |