Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00654 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015406.2 | 2 | 2820 | |||
GenBank mRNA | AK289970.1 | - | 1 | 375 | ||
AK291486.1 | - | 1 | 358 | |||
AL137476.1 | - | 1 | 1960 |