Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: LINC00320 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024090.2 | 8 | 847 | |||
NR_109786.1 | 8 | 958 | ||||
NR_109787.1 | 7 | 723 | ||||
NR_109788.1 | 7 | 784 | ||||
GenBank mRNA | AK094858.1 | - | 7 | 672 | ||
BC042856.1 | - | 8 | 802 | |||
BC044727.1 | - | 8 | 802 | |||
BC047405.1 | - | 8 | 920 | |||
BX537735.1 | - | 7 | 784 |