Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: LOC286297 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046175.1 | 4 | 1506 | |||
GenBank mRNA | AK097152.1 | - | 4 | 1508 |
Gene Symbol: LOC100133920 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024443.3 | 4 | 970 | |||
GenBank mRNA | AK074198.1 | - | 4 | 970 |
Gene Symbol: LOC100996643 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006716911.3 | XP_006716974.1 | 5 | 1047 |
Gene Symbol: LOC105379443 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011545678.2 | XP_011543980.1 | 4 | 1110 |