Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100133920 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024443.3 | 4 | 808 | |||
GenBank mRNA | AK074198.1 | - | 4 | 808 |