Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP3K14-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024434.2 | 2 | 188 | |||
NR_024435.2 | 3 | 642 | ||||
NR_110324.1 | 2 | 373 | ||||
NR_110325.1 | 4 | 368 | ||||
NR_110326.1 | 3 | 272 | ||||
GenBank mRNA | AK097219.1 | - | 2 | 1724 | ||
AK311429.1 | - | 2 | 177 | |||
AK311564.1 | - | 2 | 260 | |||
AL710937.1 | - | 4 | 339 | |||
BC031942.1 | - | 3 | 531 | |||
DB133288.1 | - | 3 | 273 | |||
DB294632.1 | - | 3 | 234 | |||
DB466340.1 | - | 2 | 372 |