Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF876P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027481.1 | 2 | 358 | |||
GenBank mRNA | BC028359.1 | - | 2 | 353 | ||
BC046475.1 | - | 2 | 329 | |||
DB024685.1 | - | 3 | 454 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI463333.1 | - | 2 | 334 | Chr. 4 - 212629 - 254157 [+] on Build GRCh38 |