Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC642361 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029407.1 | 1 | 366 | |||
GenBank mRNA | AF064843.1 | - | 1 | 362 |
Gene Symbol: LOC107985803 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017005446.1 | XP_016860935.1 | 5 | 998 |
Gene Symbol: NUTM2B-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CA312311.1 | - | 5 | 512 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CA431653.1 | - | 6 | 539 | Chr. 10 - 79797879 - 79826340 [+] on Build GRCh38 |