Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCARNA22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003004.1 | 1 | 64 | |||
GenBank mRNA | AJ609470.1 | - | 1 | 64 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV744827.1 | - | 110 | Chr. - - [-] on Build |