Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC01366 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026945.2 | 3 | 766 | |||
NR_108024.1 | 3 | 747 | ||||
NR_108025.1 | 2 | 676 | ||||
GenBank mRNA | AK093725.1 | - | 3 | 1462 | ||
BC093717.1 | - | 1 | 239 | |||
BC111994.1 | - | 1 | 239 | |||
CD518314.1 | - | 3 | 711 |