Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SOX2-OT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_004053.3 | 5 | 1514 | |||
NR_075089.1 | 4 | 1406 | ||||
NR_075090.1 | 3 | 1120 | ||||
NR_075091.1 | 8 | 1529 | ||||
NR_075092.1 | 7 | 1421 | ||||
NR_075093.1 | 5 | 1111 | ||||
GenBank mRNA | AK022826.1 | - | 5 | 1302 | ||
AL157425.1 | - | 3 | 1021 | |||
BC041898.2 | - | 5 | 1310 | |||
JN711430.1 | - | 4 | 1377 | |||
JN882275.1 | - | 3 | 1091 | |||
JQ408703.1 | - | 8 | 1528 | |||
JX110620.1 | - | 7 | 1620 | |||
JX504683.1 | - | 8 | 1582 | |||
KF981435.1 | - | 4 | 1228 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX442540.2 | - | 2 | 926 | Chr. 3 - 181699598 - 181740289 [+] on Build GRCh38 |