Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: MIR99AHG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027790.3 | 8 | 1086 | |||
NR_027791.3 | 6 | 889 | ||||
NR_111004.2 | 6 | 955 | ||||
NR_111005.2 | 7 | 1016 | ||||
NR_136542.1 | 8 | 1150 | ||||
NR_136543.1 | 5 | 828 | ||||
NR_136544.1 | 4 | 1041 | ||||
NR_136545.1 | 5 | 1156 | ||||
NR_136546.1 | 3 | 980 | ||||
NR_136547.1 | 3 | 591 | ||||
NR_136548.1 | 4 | 845 | ||||
NR_136549.1 | 4 | 772 | ||||
NR_136550.1 | 4 | 780 | ||||
NR_136551.1 | 6 | 924 | ||||
NR_136552.1 | 7 | 1039 | ||||
GenBank mRNA | AK095614.1 | - | 8 | 1086 |