Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: MIR99AHG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027790.3 | 8 | 1624 | |||
NR_027791.3 | 6 | 1427 | ||||
NR_111004.2 | 6 | 1493 | ||||
NR_111005.2 | 7 | 1554 | ||||
NR_136542.1 | 8 | 1688 | ||||
NR_136543.1 | 5 | 1366 | ||||
NR_136544.1 | 4 | 1579 | ||||
NR_136545.1 | 5 | 1694 | ||||
NR_136546.1 | 3 | 1518 | ||||
NR_136547.1 | 3 | 1129 | ||||
NR_136548.1 | 4 | 1383 | ||||
NR_136549.1 | 4 | 1310 | ||||
NR_136550.1 | 4 | 1318 | ||||
NR_136551.1 | 6 | 1462 | ||||
NR_136552.1 | 7 | 1577 | ||||
GenBank mRNA | AK095614.1 | - | 8 | 1624 |